SY3432448837 Synthonix Corporation

6-Amino-7-azaindole

  • MDL number No MDL found
  • Cas Number : 145901-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13882.html

Safety Information

MSDS https://synthonix.com/13882.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart