SY3432448868 Synthonix Corporation

5-Chloro-2-methylpyrazolo[1,5-a]pyrimidine

  • MDL number No MDL found
  • Cas Number : 189116-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13922.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/13922.html
hstatements H302 , H313 , H315 , H319 , H320 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart