SY3432448894 Synthonix Corporation

Ethyl 6-bromopyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 55899-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13951.html

Safety Information

MSDS https://synthonix.com/13951.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart