SY3432448984 Synthonix Corporation

(R)-1-Boc-2-(2-hydroxyethyl)pyrrolidine

  • MDL number MFCD20270013
  • Cas Number : 132482-06-5
  • Molecular Weight : 215.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (R)-tert-butyl 2-(2-hydroxyethyl)pyrrolidine-1-carboxylate
Inchi Code 1S/C11H21NO3/c1-11(2,3)15-10(14)12-7-4-5-9(12)6-8-13/h9,13H,4-8H2,1-3H3/t9-/m1/s1
Inchi Key MXPOLUPSEVGAAS-SECBINFHSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/14055.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/14055.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart