SY3432449001 Synthonix Corporation

Methyl 1H-imidazole-5-carboxylate

  • MDL number No MDL found
  • Cas Number : 17325-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14078.html

Safety Information

MSDS https://synthonix.com/14078.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart