SY3432449002 Synthonix Corporation

1-Methyl-1H-imidazole-4-carbonitrile

  • MDL number No MDL found
  • Cas Number : 66121-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14079.html

Safety Information

MSDS https://synthonix.com/14079.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart