SY3432449062 Synthonix Corporation

5-Aminothieno[2,3-b]pyridine

  • MDL number No MDL found
  • Cas Number : 21344-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14142.html

Safety Information

MSDS https://synthonix.com/14142.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart