SY3432449166 Synthonix Corporation

3-Iodo-1H-pyrazolo[3,4-c]pyridine

  • MDL number No MDL found
  • Cas Number : 1082040-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14261.html

Safety Information

MSDS https://synthonix.com/14261.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart