SY3432449475 Synthonix Corporation

2-(Hydroxymethyl)-3-methylpyrazine

  • MDL number No MDL found
  • Cas Number : 160818-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/14616.html

Safety Information

MSDS https://synthonix.com/14616.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart