SY3432449485 Synthonix Corporation

2-Boc-3,4-Dihydropyrrolo[1,2-a]pyrazine

  • MDL number No MDL found
  • Cas Number : 1174068-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14627.html

Safety Information

MSDS https://synthonix.com/14627.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart