SY3432449546 Synthonix Corporation

4-Chloro-2-methyl-2H-pyrazolo[3,4-b]pyridine

  • MDL number No MDL found
  • Cas Number : 944501-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/14704.html

Safety Information

MSDS https://synthonix.com/14704.html
hstatements H302 , H313 , H315 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart