SY3432449559 Synthonix Corporation

6-Chloro-4-pyridazinamine

  • MDL number No MDL found
  • Cas Number : 29049-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/14721.html

Safety Information

MSDS https://synthonix.com/14721.html
hstatements H302 , H313 , H315 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart