SY3432449592 Synthonix Corporation

1-Methyl-4-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 92534-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/14759.html

Safety Information

MSDS https://synthonix.com/14759.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart