SY3432449638 Synthonix Corporation

Ethyl 3,3-difluoro-1-methylcyclobutanecarboxylate

  • MDL number MFCD23105921
  • Cas Number : 227607-45-6
  • Molecular Weight : 178.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/14829.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/14829.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart