SY3432449872 Synthonix Corporation

1-Boc-4-hydroxypiperidine-4-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 495414-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15146.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/15146.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart