SY3432449918 Synthonix Corporation

1-(2-Methoxyethyl)-1H-pyrazole-4-boronic acid pinacol ester

  • MDL number No MDL found
  • Cas Number : 847818-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15194.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/15194.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart