SY3432449945 Synthonix Corporation

2-Chlorooxazole-4-carboxylic acid

  • Cas Number : 706789-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15228.html

Safety Information

MSDS https://synthonix.com/15228.html
Availability details for
Quantity
  • Availability
  • Add to Cart