SY3432450091 Synthonix Corporation

(R)-2-Methylmorpholine

  • MDL number No MDL found
  • Cas Number : 790184-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/15403.html

Safety Information

MSDS https://synthonix.com/15403.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart