SY3432450359 Synthonix Corporation

N-Cbz-L-4-Hydroxyproline methyl ester

  • MDL number No MDL found
  • Cas Number : 64187-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/12483.html

Safety Information

MSDS https://synthonix.com/12483.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart