SY3432450361 Synthonix Corporation

1H-Imidazo[4,5-c]pyridine-7-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 1234616-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/12557.html

Safety Information

MSDS https://synthonix.com/12557.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart