SY3432450396 Synthonix Corporation

(S)-N-Methyl-N-(piperidin-3-yl)methanesulfonamide

  • MDL number MFCD23106380
  • Cas Number : 1419075-89-0
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/13082.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/13082.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart