SY3432450412 Synthonix Corporation

3-Boc-3,7,9-triazabicyclo[3.3.1]nonane

  • MDL number No MDL found
  • Cas Number : 868407-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin China
Product Link https://synthonix.com/13119.html

Safety Information

pcodes P261 , P280 ,
MSDS https://synthonix.com/13119.html
hstatements H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart