SY3432450595 Synthonix Corporation

(1R,3S,4S)-3-Amino-4-hydroxy-cyclohexanecarboxylic acid ethyl ester hydrochloride

  • MDL number MFCD23106249
  • Cas Number : 1392745-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/13799.html

Safety Information

MSDS https://synthonix.com/13799.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart