SY3432450724 Synthonix Corporation

9-Boc-3-oxa-9-aza-bicyclo[3.3.1]nonane-7-carboxylic acid

  • MDL number MFCD22566257
  • Cas Number : 1233323-61-9
  • Molecular Weight : 271.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 9-(tert-butoxycarbonyl)-3-oxa-9-azabicyclo[3.3.1]nonane-7-carboxylic acid
Inchi Code 1S/C13H21NO5/c1-13(2,3)19-12(17)14-9-4-8(11(15)16)5-10(14)7-18-6-9/h8-10H,4-7H2,1-3H3,(H,15,16)
Inchi Key OVNHULFEMHFUNY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/14546.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/14546.html
pictograms GHS07
hstatements H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart