SY3432450946 Synthonix Corporation

(4S)-4-Mercapto-L-proline chloride

  • MDL number No MDL found
  • Cas Number : 78854-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/15630.html

Safety Information

MSDS https://synthonix.com/15630.html
hstatements H302 , H320 ,
Availability details for
Quantity
  • Availability
  • Add to Cart