SY3444790179 Synthonix Corporation

[Difluoro(phenylsulfanyl)methyl]trimethylsilane

  • MDL number MFCD19442557
  • Cas Number : 536975-49-2
  • Molecular Weight : 232.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Physical Form Solid
Shipping Temperature Ice Pak
Country of Origin US
Product Link https://synthonix.com/6812.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/6812.html
pictograms GHS05, GHS07
hstatements H302, H315, H318, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart