SY3457026283 Synthonix Corporation

5-Methoxy-3-formyl-2-(tributylstannyl)pyridine

  • MDL number MFCD32012217
  • Cas Number : 2265921-90-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/10077.html

Safety Information

signalword Danger
pcodes P261, P262, P264, P270, P273, P280
hcodes Class 6.1
MSDS https://synthonix.com/10077.html
pictograms GHS06, GHS08, GHS09
hstatements H301, H302, H312, H315, H319, H332, H335, H410
Availability details for
Quantity
  • Availability
  • Add to Cart