SY3457026292 Synthonix Corporation

2-Amino-4,6-dibromopyrimidine

  • MDL number MFCD09263251
  • Cas Number : 856973-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/1834.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/1834.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart