SY3465083935 Synthonix Corporation

1-[4-(4-amino-3-methoxyphenyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD19174778
  • Cas Number : 1021426-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/16059.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16059.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart