SY3465084004 Synthonix Corporation

5-azaspiro[2.3]hexane hemioxalate

  • MDL number MFCD27664868
  • Cas Number : 1638767-88-0
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/21417.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/21417.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart