SY3465084051 Synthonix Corporation

6-(aminomethyl)spiro[3.3]heptan-2-ol hydrochloride

  • MDL number MFCD28501613
  • Molecular Weight : 177.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/27274.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27274.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart