SY3465084054 Synthonix Corporation

exo-3-azabicyclo[3.1.0]hexan-6-ol hydrochloride

  • MDL number MFCD24843146
  • Cas Number : 1523542-00-8
  • Molecular Weight : 135.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/27611.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27611.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart