SY3465084060 Synthonix Corporation

(1R,5S,6S)-rel-8-azabicyclo[3.2.1]octan-6-ol hydrochloride

  • Cas Number : 1788041-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/27660.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/27660.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart