SY3465084066 Synthonix Corporation

cis-3-(cbz-amino)-4-hydroxypyrrolidine

  • MDL number MFCD14582423
  • Cas Number : 1262409-44-8
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/27742.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27742.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart