SY3465084119 Synthonix Corporation

tert-butyl (3S)-3-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]pyrrolidine-1-carboxylate

  • MDL number MFCD28501275
  • Cas Number : 1175273-55-8
  • Molecular Weight : 363.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/16981.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/16981.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart