SY3465084176 Synthonix Corporation

tert-butyl octahydropyrrolo[3,4-b]morpholine-6-carboxylate

  • Cas Number : 1360364-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/18989.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/18989.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart