SY3465084201 Synthonix Corporation

tert-butyl N-{[(2R)-4,4-difluoropyrrolidin-2-yl]methyl}carbamate

  • MDL number MFCD28502526
  • Cas Number : 1407991-26-7
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form oil
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/19472.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/19472.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart