SY3465084213 Synthonix Corporation

5-bromo-N-methyl-N-(1-methyl-1H-1,2,3,4-tetrazol-5-yl)pyridin-2-amine

  • MDL number MFCD28502695
  • Cas Number : 1428960-81-9
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/19679.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/19679.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart