SY3465084310 Synthonix Corporation

tert-butyl 2-formyl-6-azaspiro[3.4]octane-6-carboxylate

  • MDL number MFCD24857296
  • Cas Number : 203662-55-9
  • Molecular Weight : 239.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form liquid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/22222.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/22222.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart