SY3465084311 Synthonix Corporation

6-tert-butyl 2-methyl 6-azaspiro[3.4]octane-2,6-dicarboxylate

  • Cas Number : 203662-61-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/22224.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/22224.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart