SY3465084446 Synthonix Corporation

5-Bromo-[1,2,4]triazolo[4,3-a]pyridine

  • MDL number MFCD11043995
  • Cas Number : 1172085-67-4
  • Molecular Weight : 198.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/5059.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/5059.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart