SY3465084463 Synthonix Corporation

5-bromo-4-fluoro-1H-indazole

  • MDL number MFCD11007927
  • Cas Number : 1082041-85-7
  • Molecular Weight : 215.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/9718.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/9718.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart