SY3465084509 Synthonix Corporation

cyclobutanamine, 3-(4-methylphenoxy)-, hydrochloride (1:1), trans-

  • MDL number MFCD28166328
  • Cas Number : 1630906-70-5
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/20740.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/20740.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart