SY3465084537 Synthonix Corporation

3-chloro-4-(pyridin-2-ylmethoxy)aniline

  • MDL number MFCD08699389
  • Cas Number : 524955-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/24078.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/24078.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart