SY3465084544 Synthonix Corporation

N-(2-chloro-4-nitrophenyl)-1-hydroxynaphthalene-2-carboxamide

  • MDL number MFCD00021479
  • Cas Number : 65570-43-6
  • Molecular Weight : 342.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/24751.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/24751.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart