SY3465084624 Synthonix Corporation

1,1-dimethyl 3-oxocyclobutane-1,1-dicarboxylate

  • MDL number No MDL found
  • Cas Number : 1486409-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Country of Origin USA
Product Link https://synthonix.com/20051.html

Safety Information

pcodes P261 ,
MSDS https://synthonix.com/20051.html
hstatements H302 , H315 , H319 , H335 ,
Availability details for
Quantity
  • Availability
  • Add to Cart