SY3465084658 Synthonix Corporation

(1R,2S)-2-(3,4-difluorophenyl)cyclopropan-1-amine; (2R)-2-hydroxy-2-phenylacetic acid

  • MDL number MFCD21362980
  • Cas Number : 376608-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/23370.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/23370.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart