SY3465084668 Synthonix Corporation

2,5-dichloroquinoline

  • MDL number MFCD00832074
  • Cas Number : 59412-12-3
  • Molecular Weight : 198.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/24437.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/24437.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart