SY3465084867 Synthonix Corporation

1H-indazol-4-ylmethanamine dihydrochloride

  • MDL number MFCD28166362
  • Cas Number : 1630907-20-8
  • Molecular Weight : 220.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/27055.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/27055.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart