SY3465084886 Synthonix Corporation

3-Methyl-4H-1,2,4-triazole

  • MDL number MFCD09859189

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. Refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/15832.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/15832.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart